missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitronaphthalene 98.0+%, TCI America™
$53.90 - $232.95
Chemical Identifiers
| CAS | 86-57-7 |
|---|---|
| Molecular Formula | C10H7NO2 |
| Molecular Weight (g/mol) | 173.171 |
| MDL Number | MFCD00003913 |
| InChI Key | RJKGJBPXVHTNJL-UHFFFAOYSA-N |
| Synonym | nitrol, naphthalene, 1-nitro, alpha-nitronaphthalene, nitrol pesticide, mononitronaphthalene, naphthalene, nitro, 1-nitronaftalen, 1-nitronaphthaline, nitrol van, naphthalene, mononitro |
| PubChem CID | 6849 |
| ChEBI | CHEBI:34104 |
| IUPAC Name | 1-nitronaphthalene |
| SMILES | C1=CC=C2C(=C1)C=CC=C2[N+](=O)[O-] |
Chemical Identifiers
| 86-57-7 | |
| 173.171 | |
| RJKGJBPXVHTNJL-UHFFFAOYSA-N | |
| 6849 | |
| 1-nitronaphthalene |
| C10H7NO2 | |
| MFCD00003913 | |
| nitrol, naphthalene, 1-nitro, alpha-nitronaphthalene, nitrol pesticide, mononitronaphthalene, naphthalene, nitro, 1-nitronaftalen, 1-nitronaphthaline, nitrol van, naphthalene, mononitro | |
| CHEBI:34104 | |
| C1=CC=C2C(=C1)C=CC=C2[N+](=O)[O-] |
Specifications
| 86-57-7 | |
| Yellow | |
| MFCD00003913 | |
| 2538 | |
| RJKGJBPXVHTNJL-UHFFFAOYSA-N | |
| 1-nitronaphthalene | |
| 6849 | |
| 173.17 | |
| Crystalline Powder |
| 58°C | |
| C10H7NO2 | |
| 25 g | |
| nitrol, naphthalene, 1-nitro, alpha-nitronaphthalene, nitrol pesticide, mononitronaphthalene, naphthalene, nitro, 1-nitronaftalen, 1-nitronaphthaline, nitrol van, naphthalene, mononitro | |
| C1=CC=C2C(=C1)C=CC=C2[N+](=O)[O-] | |
| 173.171 | |
| CHEBI:34104 | |
| ≥98.0% (GC) | |
| 1-Nitronaphthalene |
Safety and Handling
EINECSNumber : (4)-0347
RTECSNumber : QJ9720000
TSCA : Yes