missing translation for 'onlineSavingsMsg'
Learn More

1-Nitronaphthalene 98.0+%, TCI America™

Catalog No. N021225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N021225G 25 g
N0212500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N021225G Supplier TCI America Supplier No. N021225G
Only null left

Chemical Identifiers

CAS 86-57-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00003913
InChI Key RJKGJBPXVHTNJL-UHFFFAOYSA-N
Synonym nitrol, naphthalene, 1-nitro, alpha-nitronaphthalene, nitrol pesticide, mononitronaphthalene, naphthalene, nitro, 1-nitronaftalen, 1-nitronaphthaline, nitrol van, naphthalene, mononitro
PubChem CID 6849
ChEBI CHEBI:34104
IUPAC Name 1-nitronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2[N+](=O)[O-]

Specifications

Melting Point 58°C
Color Yellow
Quantity 25 g
UN Number 2538
Formula Weight 173.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitronaphthalene
EINECSNumber (4)-0347
RTECSNumber QJ9720000
TSCA Yes