missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitroanthraquinone 98.0+%, TCI America™
$192.00 - $1301.11
Chemical Identifiers
| CAS | 82-34-8 |
|---|---|
| Molecular Formula | C14H7NO4 |
| Molecular Weight (g/mol) | 253.213 |
| MDL Number | MFCD00019140 |
| InChI Key | YCANAXVBJKNANM-UHFFFAOYSA-N |
| PubChem CID | 6704 |
| IUPAC Name | 1-nitroanthracene-9,10-dione |
| SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+](=O)[O-] |
Chemical Identifiers
| 82-34-8 | |
| 253.213 | |
| YCANAXVBJKNANM-UHFFFAOYSA-N | |
| 1-nitroanthracene-9,10-dione |
| C14H7NO4 | |
| MFCD00019140 | |
| 6704 | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+](=O)[O-] |
Specifications
| 82-34-8 | |
| C14H7NO4 | |
| 25 g | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+](=O)[O-] | |
| 253.213 | |
| 253.21 | |
| Crystalline Powder |
| 232°C | |
| MFCD00019140 | |
| YCANAXVBJKNANM-UHFFFAOYSA-N | |
| 1-nitroanthracene-9,10-dione | |
| 6704 | |
| ≥98.0% (GC) | |
| 1-Nitroanthraquinone |
Safety and Handling
EINECSNumber : (4)-1255
RTECSNumber : CB7940000
TSCA : Yes