missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroanthraquinone 98.0+%, TCI America™

Numéro de catalogue. N0127500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N0127500G 500 g
N012725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N0127500G Fournisseur TCI America Code fournisseur N0127500G
Il en reste null

Identifiants chimiques

CAS 82-34-8
Molecular Formula C14H7NO4
Molecular Weight (g/mol) 253.213
MDL Number MFCD00019140
InChI Key YCANAXVBJKNANM-UHFFFAOYSA-N
PubChem CID 6704
IUPAC Name 1-nitroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+](=O)[O-]

Spécifications

Melting Point 232°C
Quantity 500 g
Formula Weight 253.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitroanthraquinone
EINECSNumber (4)-1255
RTECSNumber CB7940000
TSCA Yes