missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitroanthraquinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 232°C |
| Quantity | 25 g |
| Formula Weight | 253.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Nitroanthraquinone |
Safety and Handling
| EINECSNumber | (4)-1255 |
| RTECSNumber | CB7940000 |
| TSCA | Yes |