missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitro-4-(trifluoromethoxy)benzene 96.0+%, TCI America™
$29.83 - $82.02
Chemical Identifiers
| CAS | 713-65-5 |
|---|---|
| Molecular Formula | C7H4F3NO3 |
| Molecular Weight (g/mol) | 207.11 |
| MDL Number | MFCD00040302 |
| InChI Key | UBEIKVUMDBCCRW-UHFFFAOYSA-N |
| Synonym | 1-nitro-4-trifluoromethoxy benzene, 4-trifluoromethoxy nitrobenzene, p-nitrotrifluoromethoxybenzene, 4-nitro-1-trifluoromethoxybenzene, 1-nitro-4-trifluoromethoxy-benzene, p-nitrotrifluoromethoxybenzen, benzene, 1-nitro-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4'-nitroanisole, p-nitrotrifluoroanisole |
| PubChem CID | 522405 |
| IUPAC Name | 1-nitro-4-(trifluoromethoxy)benzene |
| SMILES | [O-][N+](=O)C1=CC=C(OC(F)(F)F)C=C1 |
Chemical Identifiers
| 713-65-5 | |
| 207.11 | |
| UBEIKVUMDBCCRW-UHFFFAOYSA-N | |
| 522405 | |
| [O-][N+](=O)C1=CC=C(OC(F)(F)F)C=C1 |
| C7H4F3NO3 | |
| MFCD00040302 | |
| 1-nitro-4-trifluoromethoxy benzene, 4-trifluoromethoxy nitrobenzene, p-nitrotrifluoromethoxybenzene, 4-nitro-1-trifluoromethoxybenzene, 1-nitro-4-trifluoromethoxy-benzene, p-nitrotrifluoromethoxybenzen, benzene, 1-nitro-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4'-nitroanisole, p-nitrotrifluoroanisole | |
| 1-nitro-4-(trifluoromethoxy)benzene |
Specifications
| 713-65-5 | |
| Yellow | |
| C7H4F3NO3 | |
| 5 g | |
| UBEIKVUMDBCCRW-UHFFFAOYSA-N | |
| 1-nitro-4-(trifluoromethoxy)benzene | |
| 522405 | |
| ≥96.0% (GC) | |
| 1-Nitro-4-(trifluoromethoxy)benzene |
| 15°C | |
| 60°C | |
| MFCD00040302 | |
| 1-nitro-4-trifluoromethoxy benzene, 4-trifluoromethoxy nitrobenzene, p-nitrotrifluoromethoxybenzene, 4-nitro-1-trifluoromethoxybenzene, 1-nitro-4-trifluoromethoxy-benzene, p-nitrotrifluoromethoxybenzen, benzene, 1-nitro-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4'-nitroanisole, p-nitrotrifluoroanisole | |
| [O-][N+](=O)C1=CC=C(OC(F)(F)F)C=C1 | |
| 207.11 | |
| 207.11 | |
| Liquid |
Safety and Handling
TSCA : No