missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-4-n-propylbenzene, 96%

Catalog No. AAL0773014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0773014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0773014 Supplier Thermo Scientific Chemicals Supplier No. L0773014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10342-59-3 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10342-59-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00051839
InChI Key SXQBFCVVZIYXHV-UHFFFAOYSA-N
Synonym 1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
PubChem CID 82559
IUPAC Name 1-nitro-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point -14°C
Boiling Point 96°C to 100°C (2 mmHg)
Refractive Index 1.537
Quantity 25 g
UN Number UN2810
Beilstein 2043939
Formula Weight 165.19
Percent Purity 96%
Chemical Name or Material 1-Nitro-4-n-propylbenzene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 233-743-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only