missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-3,5-bis(trifluoromethyl)benzene 97.0+%, TCI America™

$96.94 - $318.75

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00000384
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
PubChem CID 67603
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14225G
View Documents
TCI America
B14225G
5 g
Each for $96.94
Only null left
 
B142225G
View Documents
TCI America
B142225G
25 g
Each for $318.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

328-75-6
259.11
GMUWJDVVXLBMEZ-UHFFFAOYSA-N
67603
[O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
C8H3F6NO2
MFCD00000384
1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
1-nitro-3,5-bis(trifluoromethyl)benzene

Specifications

328-75-6
72°C
MFCD00000384
1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
[O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
259.11
259.11
Liquid
Yellow
C8H3F6NO2
5 g
GMUWJDVVXLBMEZ-UHFFFAOYSA-N
1-nitro-3,5-bis(trifluoromethyl)benzene
67603
≥97.0% (GC)
1-Nitro-3,5-bis(trifluoromethyl)benzene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'