missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-3,5-bis(trifluoromethyl)benzene 97.0+%, TCI America™

Catalog No. B14225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14225G 5 g
B142225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14225G Supplier TCI America Supplier No. B14225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00000384
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
PubChem CID 67603
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
Formula Weight 259.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene