missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%

Catalog No. AAA1679714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1679714 25 g
AAA1679706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1679714 Supplier Thermo Scientific Chemicals Supplier No. A1679714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 328-75-6 | C8H3F6NO2 | 259.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00000384
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
PubChem CID 67603
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.535
Boiling Point 75°C to 76°C (11 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.427
Quantity 25 g
Beilstein 1998251
Formula Weight 259.11
Percent Purity ≥97%
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-336-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only