missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylamine, MilliporeSigma™ Supelco™

Catalog No. 111019182
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019182 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019182 Supplier Merck Emd Millipore Supplier No. 34390250MG
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 134-32-7
Density 1.114 g/mL (at 25°C)
Flash Point 157°C - Closed Cup
Physical Form Neat
Linear Formula C10H7 NH2
MDL Number MFCD00004016
Melting Point 47°C to 50°C
Boiling Point 301°C
Synonym alpha-Naphthylamine; 1-Aminonaphthalene
Type Analytical Standard
Quantity 250 mg
Formula Weight 143.19
Grade Analytical Standard
Show More Show Less