missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthylacetamide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 176°C to 178°C |
| Quantity | 25 g |
| Formula Weight | 185.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Naphthylacetamide |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | GHS P Statement P264-P270-P301+P310-P321-P405-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Specific treatment (see label). Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 201-704-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only