missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%

Catalog No. p-170997
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226890050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226890050 Supplier Thermo Scientific Chemicals Supplier No. 226890050

CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 189.0°C to 191.0°C
Color White
pH 3.5 to 4.5 (5% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C10H7OP(O)(OH)ONa·H2O
Quantity 5 g
Solubility Information (5% in water) Clear colorless to light yellow
IUPAC Name naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
Formula Weight 264.15
Percent Purity 98+%
Physical Form Fine Powder
Chemical Name or Material 1-Naphthyl phosphate, monosodium salt monohydrate, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get med
TSCA TSCA

RUO – Research Use Only