missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isothiocyanate, 98%

Catalog No. AAL1241809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1241809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1241809 Supplier Thermo Scientific Chemicals Supplier No. L1241809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 551-06-4 | C11H7NS | 185.244 g/mol

1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

So

Chemical Identifiers

CAS 551-06-4
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.244
MDL Number MFCD00003882
InChI Key JBDOSUUXMYMWQH-UHFFFAOYSA-N
Synonym 1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
PubChem CID 11080
ChEBI CHEBI:35455
IUPAC Name 1-isothiocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=S

Specifications

Melting Point 54°C to 57°C
Odor Irritating
Quantity 10 g
UN Number UN2923
Beilstein 637868
Sensitivity Moisture sensitive
Merck Index 14,6411
Solubility Information Hydrolyzes in water.
Formula Weight 185.25
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isothiocyanate
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 208-990-8
RTECSNumber NX9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only