missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isothiocyanate, 98%

Catalog No. p-170984
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128240050 5 g Glass bottle
AC128240100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128240050 Supplier Thermo Scientific Chemicals Supplier No. 128240050
Only null left

CAS: 551-06-4 | C11H7NS | 185.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 551-06-4
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.25
MDL Number MFCD00003882
InChI Key JBDOSUUXMYMWQH-UHFFFAOYSA-N
Synonym 1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
PubChem CID 11080
ChEBI CHEBI:35455
IUPAC Name 1-isothiocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.5°C to 57.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7NCS
Quantity 5 g
Beilstein 12, 1244
Fieser 01,718
Merck Index 15, 6493
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ether, benzene, hot alcohol,, acetone and ccl4
Formula Weight 185.25
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Naphthyl isothiocyanate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma symptoms
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 208-990-8
RTECSNumber NX9100000
TSCA TSCA

RUO – Research Use Only