missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthyl acetate, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 830-81-9 |
|---|---|
| Molecular Formula | C12H10O2 |
| Molecular Weight (g/mol) | 186.21 |
| MDL Number | MFCD00003922 |
| InChI Key | VGKONPUVOVVNSU-UHFFFAOYSA-N |
| Synonym | 1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate |
| PubChem CID | 13247 |
| IUPAC Name | naphthalen-1-yl acetate |
| SMILES | CC(=O)OC1=CC=CC2=CC=CC=C21 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC227250250
|
Thermo Scientific Chemicals
227250250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 830-81-9 | |
| 186.21 | |
| VGKONPUVOVVNSU-UHFFFAOYSA-N | |
| 13247 | |
| CC(=O)OC1=CC=CC2=CC=CC=C21 |
| C12H10O2 | |
| MFCD00003922 | |
| 1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate | |
| naphthalen-1-yl acetate |
Specifications
| 830-81-9 | |
| 100.0 | |
| 98.5% min. (HPLC) | |
| C12H10O2 | |
| MFCD00003922 | |
| 06, 608 | |
| VGKONPUVOVVNSU-UHFFFAOYSA-N | |
| naphthalen-1-yl acetate | |
| 13247 | |
| 99% |
| 99.0 | |
| Authentic | |
| Glass bottle | |
| CH3CO2C10H7 | |
| 25g | |
| 1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate | |
| CC(=O)OC1=CC=CC2=CC=CC=C21 | |
| 186.21 | |
| 186.21 | |
| 1-Naphthyl acetate, 99+% |
Safety and Handling
EINECSNumber : 212-599-8
RTECSNumber : QL2975500
TSCA : TSCA
RUO â Research Use Only