Learn More
1-Naphthoyl chloride, 99%
CAS: 879-18-5 | C11H7ClO | 190.63 g/mol
$97.32 - $352.71
Chemical Identifiers
| CAS | 879-18-5 |
|---|---|
| Molecular Formula | C11H7ClO |
| Molecular Weight (g/mol) | 190.63 |
| MDL Number | MFCD00004002 |
| InChI Key | NSNPSJGHTQIXDO-UHFFFAOYSA-N |
| Synonym | 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride |
| PubChem CID | 70146 |
| IUPAC Name | naphthalene-1-carbonyl chloride |
| SMILES | ClC(=O)C1=C2C=CC=CC2=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC270380100
|
Thermo Scientific Chemicals
270380100 |
10 g | Glass bottle |
N/A
|
|
||||
|
AC270380500
|
Thermo Scientific Chemicals
270380500 |
50 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 879-18-5 | |
| 190.63 | |
| NSNPSJGHTQIXDO-UHFFFAOYSA-N | |
| 70146 | |
| ClC(=O)C1=C2C=CC=CC2=CC=C1 |
| C11H7ClO | |
| MFCD00004002 | |
| 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride | |
| naphthalene-1-carbonyl chloride |
Specifications
| 879-18-5 | |
| 100.0 | |
| White | |
| 190°C (35.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.6515 to 1.6535 | |
| 10 g | |
| 1.26 | |
| Solubility in water: decomposes | |
| ClC(=O)C1=C2C=CC=CC2=CC=C1 | |
| 190.63 | |
| 190.63 | |
| Crystals, Powder or Flakes |
| 98.5 | |
| 16°C to 19°C | |
| 1.2600g/mL | |
| >110°C | |
| 98.5% min. (GC) | |
| C11H7ClO | |
| MFCD00004002 | |
| 09, 648 | |
| 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride | |
| NSNPSJGHTQIXDO-UHFFFAOYSA-N | |
| naphthalene-1-carbonyl chloride | |
| 70146 | |
| 99% | |
| 1-Naphthoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 212-903-9
RUO – Research Use Only