missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoxyacetic acid, 98+%

Catalog No. AAL0103614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0103614 25 g
AAL0103606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0103614 Supplier Thermo Scientific Chemicals Supplier No. L0103614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2976-75-2 | C12H10O3 | 202.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2976-75-2
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00003927
InChI Key GHRYSOFWKRRLMI-UHFFFAOYSA-N
Synonym 1-naphthoxyacetic acid, 1-naphthyloxy acetic acid, 2-naphthalen-1-yloxy acetic acid, 1-naphthyloxyacetic acid, naphthyloxyacetic acid, 2-1-naphthyloxy acetic acid, unii-9qi2464poa, 1-naphthoxy acetic acid, naphthalen-1-yloxy acetic acid, acetic acid, 1-naphthalenyloxy
PubChem CID 76313
ChEBI CHEBI:44588
IUPAC Name 2-naphthalen-1-yloxyacetic acid
SMILES C1=CC=C2C(=C1)C=CC=C2OCC(=O)O

Specifications

Melting Point 193°C to 197°C
Quantity 25 g
Beilstein 1912165
Formula Weight 202.21
Percent Purity ≥98%
Chemical Name or Material 1-Naphthoxyacetic acid
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 221-023-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only