missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol 99.0+%, TCI America™

Catalog No. N002625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N002625G 25 g
N0026100G 100 g
N0026500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N002625G Supplier TCI America Supplier No. N002625G

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00003930
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
PubChem CID 7005
ChEBI CHEBI:10319
IUPAC Name naphthalen-1-ol
SMILES C1=CC=C2C(=C1)C=CC=C2O

Specifications

Melting Point 96°C
Boiling Point 147°C
Quantity 25 g
UN Number 2811
Formula Weight 144.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthol
EINECSNumber (4)-0354
RTECSNumber QL2800000
TSCA Yes