missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99+%

Catalog No. p-170910
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128191000 100 g Glass bottle
AC128195000 500 g Glass bottle
AC128190025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC128191000 Supplier Thermo Scientific Chemicals Supplier No. 128191000
Only null left

1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C | CAS: 90-15-3 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
MDL Number MFCD00003930
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
PubChem CID 7005
ChEBI CHEBI:10319
IUPAC Name naphthalen-1-ol
SMILES C1=CC=C2C(=C1)C=CC=C2O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 95.0°C to 97.0°C
Color Beige-Gray to Brown
Boiling Point 278.0°C to 280.0°C
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C10H7OH
Quantity 100 g
Beilstein 06, 596
Merck Index 15, 6468
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in benzene,alcohol and ether,freely soluble in chloroform,alkali hydroxide,solutions
Formula Weight 144.17
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material 1-Naphthol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 201-969-4
RTECSNumber QL2800000
TSCA TSCA

RUO – Research Use Only