Learn More
1-Naphthol, 99%
CAS: 90-15-3 | C10H8O | 144.173 g/mol
Supplier: Thermo Scientific Chemicals A118620E
Description
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molischâs reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molischâs reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.
Solubility
Soluble in benzene, chloroform, ether and ethanol.
Notes
Air and light sensitive. Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
Specifications
| 1-Naphthol | |
| 95°C to 97°C | |
| 278°C to 280°C | |
| Weak, Phenol-like | |
| MFCD00003930 | |
| UN2811 | |
| Air and light sensitive | |
| 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern | |
| KJCVRFUGPWSIIH-UHFFFAOYSA-N | |
| naphthalen-1-ol | |
| 7005 | |
| 144.17 |
| 90-15-3 | |
| 1.28 | |
| 125°C (257°F) | |
| C10H8O | |
| 2500 g | |
| 1817321 | |
| 14,6383 | |
| Soluble in benzene,chloroform,ether and ethanol. | |
| C1=CC=C2C(=C1)C=CC=C2O | |
| 144.173 | |
| CHEBI:10319 | |
| 99% |
Chemical Identifiers
| 90-15-3 | |
| 144.173 | |
| KJCVRFUGPWSIIH-UHFFFAOYSA-N | |
| 7005 | |
| naphthalen-1-ol |
| C10H8O | |
| MFCD00003930 | |
| 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern | |
| CHEBI:10319 | |
| C1=CC=C2C(=C1)C=CC=C2O |
Safety and Handling
GHS H Statement
H318-H302-H312-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 201-969-4
RTECSNumber : QL2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only