missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99%

Catalog No. AAA1186236
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1186236 500 g
AAA1186222 100 g
AAA118620E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1186236 Supplier Thermo Scientific Chemicals Supplier No. A1186236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-15-3 | C10H8O | 144.173 g/mol

1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

Solubility
Soluble in benzene, chloroform, ether and ethanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00003930
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
PubChem CID 7005
ChEBI CHEBI:10319
IUPAC Name naphthalen-1-ol
SMILES C1=CC=C2C(=C1)C=CC=C2O

Specifications

Melting Point 95°C to 97°C
Density 1.28
Boiling Point 278°C to 280°C
Flash Point 125°C (257°F)
Odor Weak, Phenol-like
Quantity 500 g
UN Number UN2811
Beilstein 1817321
Sensitivity Air and light sensitive
Merck Index 14,6383
Solubility Information Soluble in benzene,chloroform,ether and ethanol.
Formula Weight 144.17
Percent Purity 99%
Chemical Name or Material 1-Naphthol
Show More Show Less
Hazard Category H302+H312-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H312-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-969-4
RTECSNumber QL2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only