missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99%, Spectrum™ Chemical

Catalog No. 18608129
Change view
Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608129 500 g Amber Glass Bottle
18608127 100 g Amber Glass Bottle
18608128 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608129 Supplier Spectrum Chemical Mfg Cor Supplier No. N1009500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-15-3

Specifications

CAS Max % 1
Boiling Point 288°C
Melting Range 92° to 99°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.