missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthalenemethanol 95.0+%, TCI America™
$69.31 - $275.30
Chemical Identifiers
| CAS | 4780-79-4 |
|---|---|
| Molecular Formula | C11H10O |
| Molecular Weight (g/mol) | 158.20 |
| MDL Number | MFCD00004044 |
| InChI Key | PBLNHHSDYFYZNC-UHFFFAOYSA-N |
| Synonym | 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol |
| PubChem CID | 20908 |
| ChEBI | CHEBI:38137 |
| IUPAC Name | (naphthalen-1-yl)methanol |
| SMILES | OCC1=C2C=CC=CC2=CC=C1 |
Chemical Identifiers
| 4780-79-4 | |
| 158.20 | |
| PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
| 20908 | |
| (naphthalen-1-yl)methanol |
| C11H10O | |
| MFCD00004044 | |
| 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
| CHEBI:38137 | |
| OCC1=C2C=CC=CC2=CC=C1 |
Specifications
| 4780-79-4 | |
| White-Yellow | |
| C11H10O | |
| 5 g | |
| PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
| (naphthalen-1-yl)methanol | |
| 20908 | |
| 158.20 | |
| Crystalline Powder |
| 62°C | |
| 301°C | |
| MFCD00004044 | |
| 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
| OCC1=C2C=CC=CC2=CC=C1 | |
| 158.20 | |
| CHEBI:38137 | |
| ≥95.0% (GC) | |
| 1-Naphthalenemethanol |
Safety and Handling
RTECSNumber : QJ8880000
TSCA : No