accessibility menu, dialog, popup

UserName

HO

CAS RN 4780-79-4

IUPAC Name: (naphthalen-1-yl)methanol
Synonyms: 1-naphthalenemethanol 1-naphthylenemethanol 1-menaphthyl alcohol
Molecular Weight (g/mol): 158.2
Molecular Formula: C11H10O
InChi Key: PBLNHHSDYFYZNC-UHFFFAOYSA-N
SMILES: OCC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)