missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthalenemethanol, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 4780-79-4 |
|---|---|
| Molecular Formula | C11H10O |
| Molecular Weight (g/mol) | 158.20 |
| MDL Number | MFCD00004044 |
| InChI Key | PBLNHHSDYFYZNC-UHFFFAOYSA-N |
| Synonym | 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol |
| PubChem CID | 20908 |
| ChEBI | CHEBI:38137 |
| IUPAC Name | naphthalen-1-ylmethanol |
| SMILES | OCC1=C2C=CC=CC2=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC180110020
|
Thermo Scientific Chemicals
180110020 |
2g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC180110100
|
Thermo Scientific Chemicals
180110100 |
10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4780-79-4 | |
| 158.20 | |
| PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
| 20908 | |
| naphthalen-1-ylmethanol |
| C11H10O | |
| MFCD00004044 | |
| 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
| CHEBI:38137 | |
| OCC1=C2C=CC=CC2=CC=C1 |
Specifications
| 4780-79-4 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| C10H7CH2OH | |
| 2g | |
| 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
| OCC1=C2C=CC=CC2=CC=C1 | |
| 158.20 | |
| CHEBI:38137 | |
| 98% |
| 97.5 | |
| 301°C (715.0mmHg) | |
| 98% | |
| C11H10O | |
| MFCD00004044 | |
| 06, 667 | |
| PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
| naphthalen-1-ylmethanol | |
| 20908 | |
| 158.2 | |
| 1-Naphthalenemethanol, 98% |
Safety and Handling
EINECSNumber : 225-324-1
RTECSNumber : QJ8880000
TSCA : TSCA
RUO â Research Use Only