missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethanol, 98+%

Catalog No. p-7043573
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0134709 10 g
AAL0134718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0134709 Supplier Thermo Scientific Chemicals Supplier No. L0134709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4780-79-4 | C11H10O | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4780-79-4
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.20
MDL Number MFCD00004044
InChI Key PBLNHHSDYFYZNC-UHFFFAOYSA-N
Synonym 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol
PubChem CID 20908
ChEBI CHEBI:38137
IUPAC Name naphthalen-1-ylmethanol
SMILES OCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 58°C to 63°C
Boiling Point 301°C (715 mmHg)
Quantity 10 g
Beilstein 2042532
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Naphthalenemethanol
EINECSNumber 225-324-1
RTECSNumber QJ8880000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only