missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneacetic Acid 98.0+%, TCI America™

Catalog No. p-7133568
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N000525G 25 g
N0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N000525G Supplier TCI America Supplier No. N000525G

Chemical Identifiers

CAS 86-87-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004046
InChI Key PRPINYUDVPFIRX-UHFFFAOYSA-N
Synonym 1-naphthylacetic acid, 1-naphthaleneacetic acid, 2-naphthalen-1-yl acetic acid, naphthalene-1-acetic acid, transplantone, phyomone, planofix, 1-naphthalene acetic acid, fruitone n, stop-drop
PubChem CID 6862
ChEBI CHEBI:32918
IUPAC Name 2-(naphthalen-1-yl)acetic acid
SMILES OC(=O)CC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 132°C
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthaleneacetic Acid
EINECSNumber (4)-1014
RTECSNumber QJ0875000
TSCA Yes