missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneacetic Acid, 97%, Spectrum™ Chemical

Catalog No. p-7157820
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600062 25 g Poly Bottle
18600063 100 g Poly Bottle
18600064 500 g Poly Pail
18600065 1 kg Poly Pail
18600066 5 kg Poly Pail
18600067 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600062 Supplier Spectrum Chemical Mfg Cor Supplier No. NA10725GM
Only null left

NA107, 86-87-3, C12H10O2

Spectrum™ Chemical 1-Naphthaleneacetic Acid is an organic compound that is colorless and soluble in organic solvents. It is a synthetic plant hormone that is part of the auxin family and is found in most commercial horticultural plant rooting products and is used for rooting and propagation purposes.

Chemical Identifiers

CAS 86-87-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004046
InChI Key PRPINYUDVPFIRX-UHFFFAOYSA-N
IUPAC Name 2-(naphthalen-1-yl)acetic acid
SMILES OC(=O)CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Percent Purity 97%
Grade Reagent