missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthaldehyde, 97%
Description
1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.
Solubility
Soluble in ethanol, ether, acetone, benzene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.
1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.
Solubility
Soluble in ethanol, ether, acetone, benzene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.
Specifications
Specifications
| Melting Point | 1°C to 2°C |
| Density | 1.148 |
| Boiling Point | 160°C to 161°C (15 mmHg) |
| Flash Point | 112°C (233°F) |
| Odor | Pungent |
| Refractive Index | 1.653 |
| Quantity | 50 g |
| Beilstein | 386082 |
| Sensitivity | Air sensitive; May form a precipitate |
| Solubility Information | Soluble in ethanol,ether,acetone,benzene. Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H302-H312-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 200-633-4 |
| RTECSNumber | QJ0190000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only