missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-4-iodobenzene, 98%

Catalog No. AAH2687803
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2687803 1 g
AAH2687809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2687803 Supplier Thermo Scientific Chemicals Supplier No. H2687803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62150-34-9 | C12H17I | 288.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62150-34-9
Molecular Formula C12H17I
Molecular Weight (g/mol) 288.17
MDL Number MFCD07777097
InChI Key QCTCAMDTUGSMJX-UHFFFAOYSA-N
Synonym 1-n-hexyl-4-iodobenzene, benzene, 1-hexyl-4-iodo, 4-hexylphenyl iodide
PubChem CID 15001645
IUPAC Name 1-hexyl-4-iodobenzene
SMILES CCCCCCC1=CC=C(I)C=C1

Specifications

Boiling Point 122°C to 125°C (3 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.553
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 288.16
Percent Purity 98%
Chemical Name or Material 1-n-Hexyl-4-iodobenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only