missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium trifluoromethanesulfonate, 95%, Thermo Scientific™

Catalog No. AAH5921006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5921006 5 g
AAH5921014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5921006 Supplier Thermo Scientific Chemicals Supplier No. H5921006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 460345-16-8
Molecular Formula C11H19F3N2O3S
Molecular Weight (g/mol) 316.339
MDL Number MFCD03427621
InChI Key RABFGPMWVQNDHI-UHFFFAOYSA-M
Synonym 1-hexyl-3-methylimidazolium trifluoromethanesulfonate, 1-hexyl-3-methylimidazolium trifluoromethansulfonate, 1-hexyl-3-methylimidazolium triflate, hmim otf, dsstox_cid_27847, dsstox_rid_82600, dsstox_gsid_47871, ksc491c7h, 1-hexyl-3-methylimidazoliumtrifluoromethansulfonate, 1-hexyl-3-methyl-imidazol-3-ium; trifluoromethanesulfonate
PubChem CID 2734247
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color Colorless
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 316.34
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium trifluoromethanesulfonate
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen