accessibility menu, dialog, popup

UserName

H3CNNCH3FFFSOOO

CAS RN 460345-16-8

IUPAC Name: 1-hexyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 1-hexyl-3-methylimidazolium triflate
Molecular Weight (g/mol): 316.34
Molecular Formula: C11H19F3N2O3S
InChi Key: RABFGPMWVQNDHI-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)