missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butylpyridinium bromide, 99%

Catalog No. AAH2699506
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2699506 5 g
AAH2699518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2699506 Supplier Thermo Scientific Chemicals Supplier No. H2699506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 874-80-6 | C9H14BrN | 216.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-80-6
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.12
MDL Number MFCD00671496
InChI Key KVBQNFMTEUEOCD-UHFFFAOYSA-M
Synonym 1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
PubChem CID 2734238
IUPAC Name 1-butylpyridin-1-ium;bromide
SMILES [Br-].CCCC[N+]1=CC=CC=C1

Specifications

Melting Point 105°C to 108°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 216.12
Percent Purity 99%
Chemical Name or Material 1-n-Butylpyridinium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only