missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog No. p-7021897
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1442114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1442114 Supplier Thermo Scientific Chemicals Supplier No. A1442114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20651-67-6 | Molecular Formula: C10H13I | Molecular Weight: 260.118 g/mol

It is used as an active pharmaceutical intermediate. It is also used in the synthesis and characterization of photoelectronic polymers containing triphenylamine moiety and in the palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate. It is also used in the synthesis and characterization of photoelectronic polymers containing triphenylamine moiety and in the palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.

Solubility
Sparingly soluble in water at 25°C 3.6E-3 g/L.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 20651-67-6
Molecular Formula C10H13I
Molecular Weight (g/mol) 260.118
MDL Number MFCD00051927
InChI Key XILRUONFYBUYIE-UHFFFAOYSA-N
Synonym 1-4'-iodophenyl butane, 4-n-butyliodobenzene, benzene, 1-butyl-4-iodo, 1-n-butyl-4-iodobenzene, 1-iodo-4-butylbenzene, 4-iodo-n-butylbenzene, 4-butyliodobenzene, p-iodo-n-butylbenzene, pubchem3969, 4-n-amyliodobenzene
PubChem CID 2724607
IUPAC Name 1-butyl-4-iodobenzene
SMILES CCCCC1=CC=C(C=C1)I

Specifications

Density 1.462
Boiling Point 80°C to 82°C (0.5 mmHg)
Refractive Index 1.569
Quantity 25 g
Beilstein 3030029
Sensitivity Light sensitive
Solubility Information Sparingly soluble in water at 25°C 3.6E-3g/L.
Formula Weight 260.12
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-4-iodobenzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only