missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylpyridinium tetrafluoroborate, 99%

Catalog No. AAH2698306
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2698306 5 g
AAH2698318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2698306 Supplier Thermo Scientific Chemicals Supplier No. H2698306

CAS: 597581-48-1 | C10H16BF4N | 237.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 597581-48-1
Molecular Formula C10H16BF4N
Molecular Weight (g/mol) 237.05
MDL Number MFCD08458499
InChI Key UQEAMJVKASNOMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylpyridinium tetrafluoroborate, n-butyl-3-methylpyridinium tetrafluoroborate, 1-butyl-3-methylpyridin-1-ium tetrafluoroborate, acmc-20ak5s, dsstox_cid_29281, dsstox_rid_83400, dsstox_gsid_49325
PubChem CID 17841257
IUPAC Name 1-butyl-3-methylpyridin-1-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[N+]1=CC=CC(C)=C1

Specifications

Density 1.204
Quantity 5 g
Formula Weight 237.06
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylpyridinium tetrafluoroborate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only