missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium trifluoromethanesulfonate, 98%

Catalog No. AAL1976504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976504 2 g
AAL1976509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1976504 Supplier Thermo Scientific Chemicals Supplier No. L1976504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 174899-66-2 | C9H15F3N2O3S | 288.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 174899-66-2
Molecular Formula C9H15F3N2O3S
Molecular Weight (g/mol) 288.285
MDL Number MFCD03427620
InChI Key FRZPYEHDSAQGAS-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium trifluoromethanesulfonate, 1-butyl-3-methylimidazolium trifluoromethansulfonate, 1-butyl-3-methylimidazolium triflate, c4mim triflate, bmim otf, acmc-1bwln, basionics™ vs 12, dsstox_cid_29088, dsstox_rid_83307, dsstox_gsid_49232
PubChem CID 2734246
IUPAC Name 1-butyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 16°C
Density 1.295
Flash Point None
Refractive Index 1.439
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 288.29
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium trifluoromethanesulfonate
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only