missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%

Catalog No. AAL1908709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1908709 10 g
AAL1908718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1908709 Supplier Thermo Scientific Chemicals Supplier No. L1908709

CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol

1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
PubChem CID 2734178
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Density 1.21
Flash Point None
Refractive Index 1.422
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Miscible with acetone,acetonitrile,ethyl acetate,isopropyl alcohol and methylene chloride. Immiscible with hexane,toluene and water.
IUPAC Name 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium tetrafluoroborate
Hazard Category H301-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only