Learn More
1-n-Butyl-3-methylimidazolium n-octyl sulfate, 99%
CAS: 445473-58-5 | C16H32N2O4S | 348.50 g/mol
$144.40 - $741.42
Chemical Identifiers
| CAS | 445473-58-5 |
|---|---|
| Molecular Formula | C16H32N2O4S |
| Molecular Weight (g/mol) | 348.50 |
| MDL Number | MFCD03788914 |
| InChI Key | KIDIBVPFLKLKAH-UHFFFAOYSA-M |
| Synonym | 1-butyl-3-methylimidazolium octyl sulfate, 1-butyl-3-methylimidazolium octylsulfate, 1-butyl-3-methyl-1h-imidazol-3-ium octyl sulfate, 1-butyl-3-methylimidazolium n-octyl sulfate, acmc-20aowg, bmim osu, 1-methyl-3-butylimidazolium octyl sulfate, 1-n-butyl-3-methylimidazoliumoctyl sulfate, 1-butyl-3-methylimidazolium 1-octyl sulfate, 1-n-butyl-3-methylimidazolium octyl sulfate |
| PubChem CID | 12095226 |
| IUPAC Name | 1-butyl-3-methylimidazol-3-ium;octyl sulfate |
| SMILES | CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2772006
|
Thermo Scientific Chemicals
H2772006 |
5 g |
Each for $144.40
|
|
|||||
|
AAH2772018
|
Thermo Scientific Chemicals
H2772018 |
50 g |
Each for $741.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 445473-58-5 | |
| 348.50 | |
| KIDIBVPFLKLKAH-UHFFFAOYSA-M | |
| 12095226 | |
| CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O |
| C16H32N2O4S | |
| MFCD03788914 | |
| 1-butyl-3-methylimidazolium octyl sulfate, 1-butyl-3-methylimidazolium octylsulfate, 1-butyl-3-methyl-1h-imidazol-3-ium octyl sulfate, 1-butyl-3-methylimidazolium n-octyl sulfate, acmc-20aowg, bmim osu, 1-methyl-3-butylimidazolium octyl sulfate, 1-n-butyl-3-methylimidazoliumoctyl sulfate, 1-butyl-3-methylimidazolium 1-octyl sulfate, 1-n-butyl-3-methylimidazolium octyl sulfate | |
| 1-butyl-3-methylimidazol-3-ium;octyl sulfate |
Specifications
| 445473-58-5 | |
| MFCD03788914 | |
| 1-butyl-3-methylimidazolium octyl sulfate, 1-butyl-3-methylimidazolium octylsulfate, 1-butyl-3-methyl-1h-imidazol-3-ium octyl sulfate, 1-butyl-3-methylimidazolium n-octyl sulfate, acmc-20aowg, bmim osu, 1-methyl-3-butylimidazolium octyl sulfate, 1-n-butyl-3-methylimidazoliumoctyl sulfate, 1-butyl-3-methylimidazolium 1-octyl sulfate, 1-n-butyl-3-methylimidazolium octyl sulfate | |
| CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O | |
| 348.50 | |
| 348.5 | |
| 1-n-Butyl-3-methylimidazolium n-octyl sulfate |
| C16H32N2O4S | |
| 5 g | |
| KIDIBVPFLKLKAH-UHFFFAOYSA-M | |
| 1-butyl-3-methylimidazol-3-ium;octyl sulfate | |
| 12095226 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only