accessibility menu, dialog, popup

UserName

CAS RN 445473-58-5

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium octyl sulfate
Synonyms: 1-butyl-3-methylimidazolium n-octyl sulfate
Molecular Weight (g/mol): 348.5
Molecular Formula: C16H32N2O4S
InChi Key: KIDIBVPFLKLKAH-UHFFFAOYSA-M
SMILES: CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results