missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium n-octyl sulfate, 99%

Catalog No. AAH2772018
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2772018 50 g
AAH2772006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2772018 Supplier Thermo Scientific Chemicals Supplier No. H2772018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 445473-58-5 | C16H32N2O4S | 348.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 445473-58-5
Molecular Formula C16H32N2O4S
Molecular Weight (g/mol) 348.50
MDL Number MFCD03788914
InChI Key KIDIBVPFLKLKAH-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium octyl sulfate, 1-butyl-3-methylimidazolium octylsulfate, 1-butyl-3-methyl-1h-imidazol-3-ium octyl sulfate, 1-butyl-3-methylimidazolium n-octyl sulfate, acmc-20aowg, bmim osu, 1-methyl-3-butylimidazolium octyl sulfate, 1-n-butyl-3-methylimidazoliumoctyl sulfate, 1-butyl-3-methylimidazolium 1-octyl sulfate, 1-n-butyl-3-methylimidazolium octyl sulfate
PubChem CID 12095226
IUPAC Name 1-butyl-3-methylimidazol-3-ium;octyl sulfate
SMILES CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 50 g
Formula Weight 348.5
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium n-octyl sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only