missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%

Catalog No. AAH5989318
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989318 50 g
AAH5989306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5989318 Supplier Thermo Scientific Chemicals Supplier No. H5989318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 663199-28-8 | C16H33N2O4P | 348.424 g/mol

1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

Solubility
Insoluble in water.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 663199-28-8
Molecular Formula C16H33N2O4P
Molecular Weight (g/mol) 348.424
MDL Number MFCD09265043
InChI Key NTXQJRGQUZXUMU-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium dibutyl phosphate, 1-n-butyl-3-methylimidazolium di-n-butyl phosphate, 1-butyl-3-methylimidazolium dibutylphosphate
PubChem CID 16218227
IUPAC Name 1-butyl-3-methylimidazol-3-ium;dibutyl phosphate
SMILES CCCCN1C=C[N+](=C1)C.CCCCOP(=O)([O-])OCCCC

Specifications

Color Brown
Refractive Index 1.472
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 348.42
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-n-Butyl-3-methylimidazolium di-n-butyl phosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only