CAS RN 663199-28-8
IUPAC Name:
1-butyl-3-methyl-1H-imidazol-3-ium dibutyl phosphate
Synonyms:
1-butyl-3-methylimidazolium dibutyl phosphate
Molecular Weight (g/mol):
348.42
Molecular Formula:
C16H33N2O4P
InChi Key:
NTXQJRGQUZXUMU-UHFFFAOYSA-M
SMILES:
CCCCN1C=C[N+](C)=C1.CCCCOP([O-])(=O)OCCCC
1
–
1
of
1
results
1
Promotions Available
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%
CAS: 663199-28-8 Molecular Formula: C16H33N2O4P Molecular Weight (g/mol): 348.424 MDL Number: MFCD09265043 InChI Key: NTXQJRGQUZXUMU-UHFFFAOYSA-M Synonym: 1-butyl-3-methylimidazolium dibutyl phosphate,1-n-butyl-3-methylimidazolium di-n-butyl phosphate,1-butyl-3-methylimidazolium dibutylphosphate PubChem CID: 16218227 IUPAC Name: 1-butyl-3-methylimidazol-3-ium;dibutyl phosphate SMILES: CCCCN1C=C[N+](=C1)C.CCCCOP(=O)([O-])OCCCC