missing translation for 'onlineSavingsMsg'
Learn More

1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene, 90%

Catalog No. p-3771256
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426900010 1 g Glass bottle
AC426900050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426900010 Supplier Thermo Scientific Chemicals Supplier No. 426900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108468-00-4 | C13H20N2O2 | 236.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108468-00-4
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.31
MDL Number MFCD02683058
InChI Key NUANLVJLUYWSER-UHFFFAOYSA-N
Synonym 1-n-boc-aminomethyl-4-aminomethyl benzene, tert-butyl 4-aminomethyl benzylcarbamate, tert-butyl n-4-aminomethyl benzyl carbamate, 4-aminomethyl-benzyl-carbamic acid tert-butyl ester, tert-butyl n-4-aminomethyl phenyl methyl carbamate, n-4-aminomethyl phenyl methyl tert-butoxy carboxamide, n-boc-p-xylylenediamine, 4-tert-butoxycarbonylaminomethyl benzylamine
PubChem CID 3354775
IUPAC Name tert-butyl N-[[4-(aminomethyl)phenyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 64.0°C to 68.0°C
Color Beige or Pink-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H4CH2NHCO2C(CH3)3
Quantity 1 g
Formula Weight 236.31
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

RUO – Research Use Only