missing translation for 'onlineSavingsMsg'
Learn More

1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane 96.0+%, TCI America™

Catalog No. h036125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H036125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H036125G Supplier TCI America Supplier No. H036125G
Only null left

Chemical Identifiers

CAS 14712-23-3
Molecular Formula C15H30N2O
Molecular Weight (g/mol) 254.418
MDL Number MFCD00023766
InChI Key KTRJPKYFFZFQJB-UHFFFAOYSA-N
PubChem CID 84604
IUPAC Name 2-[4-(3-piperidin-4-ylpropyl)piperidin-1-yl]ethanol
SMILES C1CNCCC1CCCC2CCN(CC2)CCO

Specifications

Melting Point 83°C
Color White
Boiling Point 250°C
Quantity 25 g
Formula Weight 254.42
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[N-(2-Hydroxyethyl)-4′-piperidyl]-3-(4′-piperidyl)propane