accessibility menu, dialog, popup

UserName

CAS RN 14712-23-3

OHNNH

CAS RN 14712-23-3

IUPAC Name: 2-{4-[3-(piperidin-4-yl)propyl]piperidin-1-yl}ethan-1-ol
Synonyms: 2-{4-[3-(piperidin-4-yl)propyl]piperidin-1-yl}ethanol
Molecular Weight (g/mol): 254.42
Molecular Formula: C15H30N2O
InChi Key: KTRJPKYFFZFQJB-UHFFFAOYSA-N
SMILES: OCCN1CCC(CCCC2CCNCC2)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results