missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Morpholino-1-cyclohexene 97.0+%, TCI America™
Supplier: TCI America C0956500ML
Specifications
| 1-Morpholino-1-cyclohexene | |
| Yellow | |
| C10H17NO | |
| 500 mL | |
| IIQFBBQJYPGOHJ-UHFFFAOYSA-N | |
| 4-(cyclohexen-1-yl)morpholine | |
| 69589 | |
| ≥97.0% (GC,T) |
| 670-80-4 | |
| 140°C | |
| MFCD00006163 | |
| 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene | |
| C1CCC(=CC1)N2CCOCC2 | |
| 167.252 | |
| 167.25 | |
| Liquid |
Chemical Identifiers
| 670-80-4 | |
| 167.252 | |
| IIQFBBQJYPGOHJ-UHFFFAOYSA-N | |
| 69589 | |
| C1CCC(=CC1)N2CCOCC2 |
| C10H17NO | |
| MFCD00006163 | |
| 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene | |
| 4-(cyclohexen-1-yl)morpholine |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator