missing translation for 'onlineSavingsMsg'
Learn More

1-Morpholino-1-cyclohexene 97.0+%, TCI America™

Catalog No. p-7124705
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C095625ML 25 mL
C0956500ML 500 mL
2 options

Catalog No. C095625ML

Supplier: TCI America C095625ML

Only null left

Chemical Identifiers

CAS 670-80-4
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00006163
InChI Key IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Synonym 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene
PubChem CID 69589
IUPAC Name 4-(cyclohexen-1-yl)morpholine
SMILES C1CCC(=CC1)N2CCOCC2

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 mL
Formula Weight 167.25
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Morpholino-1-cyclohexene
TSCA Yes
Recommended Storage Refrigerator