missing translation for 'onlineSavingsMsg'
Learn More

1-Morpholino-1-cyclododecene 95.0+%, TCI America™

Catalog No. M19395G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19395G Supplier TCI America Supplier No. M19395G
Only null left

Chemical Identifiers

CAS 3725-39-1
Molecular Formula C16H29NO
Molecular Weight (g/mol) 251.414
MDL Number MFCD00042788
InChI Key SZTBSJOMNRSBGA-MHWRWJLKSA-N
Synonym 4-(1-Cyclododecen-1-yl)morpholine
PubChem CID 5371595
IUPAC Name 4-[(1E)-cyclododecen-1-yl]morpholine
SMILES C1CCCCCC(=CCCCC1)N2CCOCC2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 251.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Morpholino-1-cyclododecene
TSCA No
Recommended Storage Freezer