accessibility menu, dialog, popup

UserName

NO(E)

CAS RN 3725-39-1

IUPAC Name: 4-[(1E)-cyclododec-1-en-1-yl]morpholine
Synonyms: 4-[(1E)-cyclododec-1-en-1-yl]morpholine
Molecular Weight (g/mol): 251.41
Molecular Formula: C16H29NO
InChi Key: SZTBSJOMNRSBGA-MHWRWJLKSA-N
SMILES: C1CN(CCO1)C1=C/CCCCCCCCCC\1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results